Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Hepatotox Set™ 1
Hepatotox Set™ 1
Hepatotox Set™ 1
Hepatotox Set™ 1
Catalog # ENZOALX850325KI01
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Hepatotox Set™ 1
Catalog # ENZOALX850325KI01
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 SET
Seq:
0125
Specifications
About this item
Product Details
:
Product Details