

Ryania ≥99% (by HPLC), high purity
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:15662-33-6
:
Formula:C₂₅H₃₅NO₉
:493.553 g/mol
Merck Index:14,08307
Storage Temperature:Freezer
EINECS:239-732-2
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Source:Isolated from Ryania speciosa, Flacourtiaceae
- Purity:≥99% (HPLC)