X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:7240-90-6
:
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666
:408.63 g/mol
Melting Pt:237…239 °C
Storage Temperature:Freezer
EINECS:230-640-8