

Lipopolysaccharides 1 mg/ml in water (from S. enteritidis) S-Form, TLRgrade™ ready-to-use
Catalog # ENZOALX581019L002
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:2 mL
- Seq:2145
Specifications
Specification Test Results
- Concentration:1 mg/ml
- Formulation:Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
- Purity:Absence of detectable protein or DNA contaminants with agonistic TLR activity
Specification Test Results
About this item
:
:GHS08
:GHS07
Storage Temperature:Refrigerator