Order Entry
Germany
ContactUsLinkComponent
 
2-Phosphonomethyl pentanedioic acid ≥97% (by HPLC)
Catalog # ENZOALX550358M005
Supplier:  Enzo Life Sciences
CAS Number:  
2-Phosphonomethyl pentanedioic acid ≥97% (by HPLC)
Catalog # ENZOALX550358M005
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :226.12 g/mol
Storage Temperature:Freezer
CAS Number:173039-10-6
Formula:C₆H₁₁O₇P
MDL Number:MFCD00940167