Order Entry
Germany
ContactUsLinkComponent
Nicardipine hydrochloride ≥98%
Nicardipine hydrochloride ≥98%
Catalog # ENZOALX550273G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Nicardipine hydrochloride ≥98%
Catalog # ENZOALX550273G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :515.99 g/mol
CAS Number:54527-84-3
Formula:C₂₆H₂₉N₃O₆
MDL Number:MFCD00057327