Order Entry
Germany
Orders LinkContactUsLinkComponent
 

D-(+)-Biotin

Supplier:   Enzo Life Sciences
D-(+)-Biotin
Supplier:  Enzo Life Sciences
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:13,01231
  • Melting Pt:232…233 °C
  • Density:1,41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 52,80 €
    52,80 €
    1 * 1 g
       52,80 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 178,00 €
    178,00 €
    1 * 5 g
       178,00 €