Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Carboxy-PTIO potassium salt
Carboxy-PTIO potassium salt ≥99% (by HPLC)
Carboxy-PTIO potassium salt ≥99% (by HPLC)
:
ENZO LIFE SCIENCES
Carboxy-PTIO potassium salt ≥99% (by HPLC)
:
ENZO LIFE SCIENCES
:
148819-94-7
:
:
C₁₄H₁₆KN₂O₄
:
MFCD00216153
:
315.4 g/mol
:
Freezer