

Gardiquimod ≥98% (by HPLC and UV method)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:1020412-43-4
:
Formula:C₁₇H₂₃N₅O
:313.4 g/mol
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Purity:≥98% (HPLC (UV))