Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Mitoxantrone dihydrochloride
Mitoxantrone dihydrochloride (synthetic) ≥97%
Mitoxantrone dihydrochloride (synthetic) ≥97%
Mitoxantrone dihydrochloride (synthetic) ≥97%
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Mitoxantrone dihydrochloride (synthetic) ≥97%
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
70476-82-3
:
Formula:
C₂₂H₃₀Cl₂N₄O₆
MDL Number:
MFCD00242943
:
517.404 g/mol
Storage Temperature:
Freezer