Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Platin
cis-Platin
cis-Platin
cis-Platin
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
cis-Platin
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
15663-27-1
:
Formula:
H₆Cl₂N₂Pt
MDL Number:
MFCD00011623
:
300.05 g/mol
Merck Index:
13,02341
Density:
3,738 g/cm³ (20 °C)
EINECS:
239-733-8
UN:
3288
ADR:
6.1,II