Order Entry
Germany
ContactUsLinkComponent
 
Delphinidin chloride ≥97% (by HPLC)
Catalog # ENZOALX385028M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Delphinidin chloride ≥97% (by HPLC)
Catalog # ENZOALX385028M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :338.71 g/mol
Storage Temperature:Freezer
CAS Number:528-53-0
Formula:C₁₅H₁₁O₇Cl
MDL Number:MFCD00016663