Order Entry
Germany
ContactUsLinkComponent
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥98% (by HPLC)
Irritant
Health
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥98% (by HPLC)
Catalog # ENZOALX385024M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
Health
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone ≥98% (by HPLC)
Catalog # ENZOALX385024M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Identity:
    Determined by NMR and MS

Specification Test Results

About this item

 :
 :316.27 g/mol
EINECS:207-545-5
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:480-19-3
Formula:C₁₆H₁₂O₇
MDL Number:MFCD00017310