Amikacin disulphate
Catalog # ENZOALX380266G001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:781.77 g/mol
Storage Temperature:Freezer
CAS Number:39831-55-5
Formula:C₂₂H₄₃N₅O₁₃·2H₂SO₄
MDL Number:MFCD00167475