Order Entry
Germany
ContactUsLinkComponent
 
Bafilomycin B1 (from Streptomyces griseus) ≥95% (by HPLC)
  ENZOALX380063M001
 :  
Bafilomycin B1 (from Streptomyces griseus) ≥95% (by HPLC)
  ENZOALX380063M001
 :  

 

  • Pk:
    1 mg
  • Seq:
    2070

 

 

 :
 :815.99 g/mol
 :Freezer
 :88899-56-3
 :80112-34-1
 :C₄₄H₆₅NO₁₃
 :MFCD00242671