Order Entry
Germany
ContactUsLinkComponent
Galiellalactone (from unidentified fungus MST-FP1888) ≥95% (by HPLC)
Galiellalactone (from unidentified fungus MST-FP1888) ≥95% (by HPLC)
Catalog # ENZOALX350336MC05
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Galiellalactone (from unidentified fungus MST-FP1888) ≥95% (by HPLC)
Catalog # ENZOALX350336MC05
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Seq:
    2035

Specifications

About this item

 :
 :194.23 g/mol
Storage Temperature:Freezer
CAS Number:133613-71-5
Formula:C₁₁H₁₄O₃