Order Entry
Germany
ContactUsLinkComponent
R-(-)-Glucoraphanin potassium salt ≥99% (by HPLC)
R-(-)-Glucoraphanin potassium salt ≥99% (by HPLC)
Catalog # ENZOALX350181M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
R-(-)-Glucoraphanin potassium salt ≥99% (by HPLC)
Catalog # ENZOALX350181M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :475.58 g/mol (anhy)
CAS Number:21414-41-5
MDL Number:MFCD08460269