SMI-4a ≥95%
Catalog # ENZOALX270515M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:273.23 g/mol
CAS Number:327033-36-3
:438190-29-5
Formula:C₁₁H₆F₃NO₂S
MDL Number:MFCD01152003