Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Midostaurin
Midostaurin ≥98% (by TLC)
Midostaurin ≥98% (by TLC)
Midostaurin ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Midostaurin ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
120685-11-2
:
MFCD00871372
Formula:
C₃₅H₃₀N₄O₄
MDL Number:
MFCD12828879
:
570.64 g/mol
Storage Temperature:
Freezer