Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Mitomycin C ≥95%

Supplier:   Enzo Life Sciences
Mitomycin C ≥95%
Supplier:  Enzo Life Sciences
Certificates
Mitomycin C ≥95%
Mitomycin C ≥95%

2 Options Available Below

Product Details & Documents

Antitumor antibiotic. DNA crosslinking and damaging agent. Inhibitor of DNA synthesis, nuclear division and cancer cells. DNA intercalating agent. Induces apoptosis in a variety of cells via a p53-dependent pathway.

  • Blue-violet solid

Soluble in water (1 mg/mL, warm), 100% ethanol (1 mg/mL, warm), DMSO (15 mg/mL) or methanol

Synonyms:  Mitomycin, Ametycin
  • CAS Number:50-07-7
  • Formula:C₁₅H₁₈N₄O₅
  • MDL Number:MFCD00078109
  • Molecular Weight:334.33 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 2 mg
    Log in for availability
    Log in for availability
    1 * 2 mg
    1 * 2 mg - 64,90 €
    64,90 €
    1 * 2 mg
       64,90 €

  • 10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 209,00 €
    209,00 €
    1 * 10 mg
       209,00 €