Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Puromycin dihydrochloride
Puromycin dihydrochloride ≥98% (by HPLC)
Puromycin dihydrochloride ≥98% (by HPLC)
:
ENZO LIFE SCIENCES
Puromycin dihydrochloride ≥98% (by HPLC)
:
ENZO LIFE SCIENCES
:
58-58-2
:
:
C₂₂H₂₉N₇O₅·2HCl
:
MFCD00012691
:
544.44 g/mol
:
Freezer