Argatroban ≥98% (by HPLC)
Catalog # ENZOBMLPI1460010
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:508.64 g/mol
Storage Temperature:Freezer
CAS Number:74863-84-6
:141396-28-3
Formula:C₂₃H₃₆N₆O₅S
MDL Number:MFCD23102419