Order Entry
Germany
ContactUsLinkComponent
AC-55469 ≥98% (by HPLC)
AC-55469 ≥98% (by HPLC)
Catalog # ENZOBMLGR1120010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
AC-55469 ≥98% (by HPLC)
Catalog # ENZOBMLGR1120010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :310.4 g/mol
Storage Temperature:Ambient
CAS Number:59662-49-6
MDL Number:MFCD00157040