Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Auftragserfassung
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FCCP
FCCP ≥98% (durch TLC)
FCCP ≥98% (durch TLC)
FCCP ≥98% (durch TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
FCCP ≥98% (durch TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
370-86-5
:
Formula:
C₁₀H₅F₃N₄O
MDL Number:
MFCD00009699
:
254.17 g/mol