Order Entry
Germany
ContactUsLinkComponent
Ingenol-3,20-dibenzoate ≥97%
Irritant
Health
Ingenol-3,20-dibenzoate ≥97%
Catalog # ENZOBMLPE1860001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
Health
Ingenol-3,20-dibenzoate ≥97%
Catalog # ENZOBMLPE1860001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Purity:
    ≥97%

Specification Test Results

About this item

 :
 :556.7 g/mol
 :GHS07
 :GHS08
Storage Temperature:Freezer
CAS Number:59086-90-7
Formula:C₃₄H₃₆O₇