Order Entry
Germany
ContactUsLinkComponent
N-Desmethyl clozapine ≥99% (by HPLC), yellow solid
Irritant
N-Desmethyl clozapine ≥99% (by HPLC), yellow solid
Catalog # ENZOBMLNS1060025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Irritant
N-Desmethyl clozapine ≥99% (by HPLC), yellow solid
Catalog # ENZOBMLNS1060025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥99% (HPLC)
  • Identity:
    Determined by ¹H-NMR

Specification Test Results

About this item

 :
 :312.8 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:6104-71-8
Formula:C₁₇H₁₇ClN₄
MDL Number:MFCD00210189