Order Entry
Germany
ContactUsLinkComponent
Phosphatidic acid, dioleoyl ≥98% (by GC), white powder
Health
Phosphatidic acid, dioleoyl ≥98% (by GC), white powder
Artikelnr. ENZOBMLLP1040010
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Health
Phosphatidic acid, dioleoyl ≥98% (by GC), white powder
Artikelnr. ENZOBMLLP1040010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Purity:
    ≥98% (GC)

Specification Test Results

About this item

 :
 :724 g/mol
 :GHS08
Storage Temperature:Freezer
CAS Number:108392-02-5
Formula:C₃₉H₇₃O₈P·Na
MDL Number:MFCD00674303