Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phaclofen
Phaclofen ≥98% (by HPLC), white solid
Phaclofen ≥98% (by HPLC), white solid
ENZOBMLEA1270025
:
ENZO LIFE SCIENCES
:
Phaclofen ≥98% (by HPLC), white solid
ENZOBMLEA1270025
:
ENZO LIFE SCIENCES
:
Pk:
25 mg
Seq:
2315
Identity:
Determined by H-NMR and MS
Purity (HPLC):
≥98%
:
:
249.6 g/mol
:
GHS07
:
Ambient
:
114012-12-3
:
C₉H₁₃ClNO₃P