Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,9-Pyrazoloanthrone
1,9-Pyrazoloanthrone ≥98% (by TLC)
1,9-Pyrazoloanthrone ≥98% (by TLC)
1,9-Pyrazoloanthrone ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
1,9-Pyrazoloanthrone ≥98% (by TLC)
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
129-56-6
:
Formula:
C₁₄H₈N₂O
MDL Number:
MFCD00022289
:
220.231 g/mol
Storage Temperature:
Ambient