Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
BAY 11-7085
BAY 11-7085 ≥99%
BAY 11-7085 ≥99%
BAY 11-7085 ≥99%
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
BAY 11-7085 ≥99%
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
196309-76-9
:
Formula:
C₁₃H₁₅NO₂S
MDL Number:
MFCD01862602
:
249.328 g/mol