Order Entry
Germany
ContactUsLinkComponent
(±)-Propranolol hydrochloride ≥98% (by HPLC)
(±)-Propranolol hydrochloride ≥98% (by HPLC)
Catalog # ENZOBMLST4050005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
(±)-Propranolol hydrochloride ≥98% (by HPLC)
Catalog # ENZOBMLST4050005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :295.81 g/mol
Storage Temperature:Freezer
CAS Number:318-98-9
 :3506-09-0
Formula:C₁₆H₂₁NO₂·HCl
MDL Number:MFCD00012558