Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nafamostat mesylate
Nafamostat mesylate ≥98%
Nafamostat mesylate ≥98%
Nafamostat mesylate ≥98%
Supplier:
ENZO LIFE SCIENCES
Certificates
Expand image
Nafamostat mesylate ≥98%
Supplier:
ENZO LIFE SCIENCES
About this item
CAS Number:
82956-11-4
:
Formula:
C₁₉H₁₇N₅O₂
MDL Number:
MFCD00941430
:
539.58 g/mol