

Clozapine ≥99% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:5786-21-0
:
Formula:C₁₈H₁₉ClN₄
MDL Number:MFCD00153785
:326.83 g/mol
Merck Index:14,02423
Storage Temperature:Refrigerator
EINECS:227-313-7
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Purity:≥99% (HPLC)
- Identity:Determined by IR