Nicotinamide
500 µl
Catalog # ENZOBMLKI2830500
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Nicotinamide, the water-soluble amide of nicotinic acid, is a component of the two most important coenzymes-nicotinamide adenine dinucleotide and nicotinamide adenine dinucleotide phosphate. Nicotinamide is involved in numerous oxidation-reduction reactions in mammalian biological systems acting as an antioxidant.
Soluble in water, 100% ethanol or glycerol. Slightly soluble in chloroform, almost insoluble in ether or acetone.
Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
- CAS Number:98-92-0
- Formula:C₆H₆N₂O
- MDL Number:MFCD00006395
- Molecular Weight:122.13 g/mol
- Merck Index:14,06523
- Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
- EINECS:202-713-4
Specifications
Specification Test Results
- Formulation:50 mM nicotinamide in 50 mM tris/Cl, pH 8.0, 137 mM NaCl, 2.7 mM KCl, 1 mM MgCl₂.
Specifications
- Pk:500 µl
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



