Order Entry
Germany
ContactUsLinkComponent
Resveratrol ≥98% (by TLC)
Resveratrol ≥98% (by TLC)
Catalog # ENZOBMLFR1040100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Resveratrol ≥98% (by TLC)
Catalog # ENZOBMLFR1040100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :228.25 g/mol
CAS Number:501-36-0
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00133799