ExoBrite™ EV Surface Stain Sampler Kit
Catalog # 30127.
Supplier: Biotium
New Product
CAS Number:
Specifications
- Pk:1
- Colour:Green
- Description:ExoBrite™ EV Surface stain sampler kit
Specifications
About this item
The ExoBrite™ EV Surface Stain Sampler Kit, Green was developed to offer each of Biotium’s ExoBrite™ EV Surface Stains (CTB, WGA, and Annexin V) for assessing which stain offers the best coverage for the EV samples of interest.