ContactUsLinkComponent
Standard Screw Caps for Microtubes
  SORE390751CL
New Product
Standard Screw Caps for Microtubes
  SORE390751CL
New Product
  • Closure type:
    Screw cap
  • Colour:
    Red
  • Pk:
    1.000
Made of virgin polyethylene; medical grade.
  • Free of DNase, RNase and endotoxin
  • With O-ring and without seal
  • Non sterile
CE IVD marking.