Order Entry
Germany
ContactUsLinkComponent
Garmet Hanger, Closed Hook
Garmet Hanger, Closed Hook
Catalog # TEKMCRGS-GHC
Supplier:  TEKNOMEK
Garmet Hanger, Closed Hook
Catalog # TEKMCRGS-GHC
Supplier:  TEKNOMEK

Specifications

  • Description:
    Closed hook garment hanger, 380W×200H mm
  • Pk:
    1

Specifications

About this item

Closed hook garment hanger are manufactured from fully welded 304-grade stainless steel with a polished finish.

  • Ideal addition to hooked garment rails

Weight: 0,220 kg.