Copper indium sulphide zinc sulphide (CuInS2/ZnS) 1 mg/ml in toluene, core-shell type quantum dots fluorescence λem 650 nm, Sigma-Aldrich®
Catalog # SIAL924326-1MG
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- fluorescence:FWHM <125 nm - λem 650 nm, quantum yield ≥40%
Specification Test Results
About this item
:
UN:1294
:GHS02
:GHS07
:GHS08
Storage Temperature:Refrigerator
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:4.4 °C (39.9 °F)