Tablet reagents for Chlorine Combined and Total (Extended Range DPD XF and XT)
Catalog # PALIPM033
Supplier: Palintest
CAS Number:
Specifications
- Pk:50
- Description:Tablet reagent for chlorine – free, combined and total (extended range DPD XF and XT)
- Environmentally Preferable:
- Measurement range:0 - 10 mg/L Cl₂
- Tests per kit:50
Specifications
About this item
Tablet reagents for chlorine – free, combined and total (extended range DPD XF and XT).
Ordering information: Starter Packs (PM codes) contain 50 tests plus any optional reagents or equipment needed.