Order Entry
Germany
ContactUsLinkComponent
 
Laser Protection Goggles
Catalog # UNVT562S.00.01.312
Supplier:  UNIVET
Laser Protection Goggles
Catalog # UNVT562S.00.01.312
Supplier:  UNIVET

Specifications

  • Colour:
    Pink
  • Description:
    Laser saftey goggles, UL-1008, polycarbonate
  • Pk:
    1

Specifications

About this item

  • Filter: Full protection
  • VLT: 40%
  • Alignement laser wavelength T%(λ) >10%: 410 to 695 nm