ContactUsLinkComponent
VHG™ Optilene-XF™ XRF Thin Film
VHG™ Optilene-XF™ XRF Thin Film
Catalog # PROOVHG-FOL06-R3
VHG™ Optilene-XF™ XRF Thin Film
Catalog # PROOVHG-FOL06-R3
Specifications
  • Width:
    76,2 mm
  • Description:
    Optilene-XF™ XRF thin film
  • Length:
    91,44 m
  • Thickness:
    6 μm
  • Pk:
    1 Roll
Specifications
About this item
Thin films for XRF sample cups.
Highest grade, proprietary film featuring optimal transmission, purity, strength, chemical resistance, and thermal stability. Good for full range of XRF analytes, including light elements.