Order Entry
Germany
ContactUsLinkComponent
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine 95%
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine 95%
Catalog # SIAL537284-SPEC
Supplier:  Merck
CAS Number:  
N-(3-sulphopropyl)-N-methacroyloxyethyl-N,N-dimethylammonium betaine 95%
Catalog # SIAL537284-SPEC
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 kg
  • Seq:
    4158

Specifications

Specification Test Results

  • Appearance (Colour):
    White
  • Appearance (Form):
    Powder or Crystals
  • Infrared Spectrum:
    Conforms to Structure
  • Carbon Content:
    44.7 - 47.3%
  • Hydrogen Content:
    7.2 - 8.0%
  • Nitrogen Content:
    4.7 - 5.0%
  • Sulphur Content:
    10.9 - 11.5%
  • Solubility (Turbidity):
    Clear
  • Solubility (Colour) 25 mg/ml, H₂O:
    Colourless

Specification Test Results

About this item

 :
 :279.351 g/mol
Melting Pt:150...155 °C (lit.)
 :M-Clarity™ quality level = MQ200
CAS Number:3637-26-1
Formula:C₁₁H₂₁NO₅S
MDL Number:MFCD00040574