Order Entry
Germany
Orders LinkContactUsLinkComponent
Gas Washing Bottle

Gas Washing Bottle

Muencke insert for 30 - 100 ml bottle, 12 mm, 2× GL 14
Catalog # NEUB0578-60-121414
Gas Washing Bottle
Catalog # NEUB0578-60-121414
Gas Washing Bottle

Muencke insert for 30 - 100 ml bottle, 12 mm, 2× GL 14

Price
88,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Components individually, from Boro 3.3; inserts with safety glass ring.

Specifications
  • Ext.Ø:
    12 mm
  • Thread:
    2× GL 14
  • Description:
    Muencke insert for 30 - 100 ml bottle
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.