ContactUsLinkComponent
Rechargeable NiMH Battery for M510/M610/M710 Printers
Rechargeable NiMH Battery for M510/M610/M710 Printers
Catalog # BRDY175674
Supplier:  Brady
undefined
Rechargeable NiMH Battery for M510/M610/M710 Printers
Catalog # BRDY175674
Supplier:  Brady
Specifications
  • Description:
    Rechargeable NiMH battery for M510/M610/M710 printers, grey, 57,00W×109,98H×25,00D mm
  • Pk:
    1
Specifications
About this item
Work longer with this NiMH M510, M610 and M710 battery pack. It enables cordless printer operation so you can print labels where (and when) you need them.
  • Removable, rechargeable Nickel-metal hydride battery pack
  • Battery can be charged only while installed in an M510, M610 or M710 printer
  • Prints up to 1,700 labels per charge
Work longer with this NiMH M510, M610 and M710 Battery Pack. It enables cordless printer operation so you can print labels where (and when) you need them. Removable, rechargeable Nickel-metal Hydride Battery Pack. Battery can be charged only while installed in an M510, M610 or M710 printer. Prints up to 1,700 labels per charge.
Ordering information: Order reference M-NIMH-BATTERY.
Delivery: NiMH battery pack.