Anti-GLUT8 Rabbit Polyclonal Antibody
Catalog # BSBTPA2165-10UG
Supplier: Boster Bio
New Product
Specifications
- Antibody type:Primary
- Antigen name:Glucose Transporter 8
- Antigen symbol:GLUT8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Near the C-terminus
- Cross adsorption:No
- Form:Lyophilised
- Amino acid number:461 - 477
- Sequence:ETKGRTLEQVTAHFEGR
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse Glucose Transporter 8 (461-477aa ETKGRTLEQVTAHFEGR), different from the related rat sequence by one amino acid, and from the related human sequence by two amino acids.
- Purification:Purified by immunogen affinity purification
- Pk:10 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Solute carrier family 2, facilitated glucose transporter member 8 (SLC2A8) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 μg/ml.
Type: Primary
Antigen: GLUT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat