Ficoll®, MP Biomedicals
Catalog # ICNA0216000380
Supplier: MP Biomedicals
Specifications
- Description:Ficoll®
- Pk:100 g
Specifications
About this item
A copolymer of sucrose and epichlorohydrin. Component used to make density gradients for lymphocyte separation.