VWR® Hitachi UH4150 UV-VIS/NIR Spectrophotometers
These UV-Vis/NIR spectrophotometers offer enormous flexibility for the evaluation and quality control of solid samples, typically optical components or coated surfaces. They can be configured for non destructive measurements when equipped with relevant accessories for applications such as:
- Micro sample reflectance
- Wafer reflectance/transmission
- Optical thin film reflection
- Lens transmittance
- Solar cells and coated glass evaluation
High precision accessories can measure at a range of incident angles.
For coated glass surfaces, the variable angle absolute reflectance accessory (20 to 60°) measures the absolute reflectance and transmittance at a desired angle by rotating the detector and the sample stage independently.
: For more information on the range of accessories, please contact your local Avantor agency.
- Catalog No:
- 634-0952
- 634-0951
- Wavelength range:
- 240 - 2600 nm
- 240 - 2600 nm
- Monochromator:
- Prism/grating infinitely variable slit and integrating sphere
- Prism/grating infinitely variable slit and integrating sphere
- Sample size:
- 430×430 mm
- 430×430 mm