ExGuard ER600W Temperature Controlled Storage Cabinet
Catalog # GRAM986310046
Supplier: GRAM
Specifications
- W×D×H ext.:820×789×1996/2246 mm
- Noise level:46,3 dB(A)
- Model:ExGuard ER600W
- Electrical:230 V, 50 Hz, 16 A, 316 W
- Refrigerant:R290
- Access ports:1 pcs, Ø 24,5 mm
- Alarm:Acoustic and visual, high/low temperature alarms
- Defrost:Automatic smart defrost with re-evaporation of defrost water
- Door type:Solid/glass door
- Shelves/drawers:47 shelves, 9 drawers
- Weight:Unpackaged net weight without optional fittings: 166 kg
- Insulation:70 mm polyurethane with HFC-free cyclopentane propellant
- Pk:1
- Temperature:−2/+20 °C
Specifications
About this item
The ExGuard is designed for safe temperature controlled storage of items potentially emitting noxious, malodourous and/or explosive atmospheres. At the core, the ExGuard is envisioned to facilitate a safer work environment for users.
- Built in safety mechanisms that prevent the cabinet from being opened before the hazardous atmosphere have been safely extracted
- Effortless and safe daily operation
- ATEX Cat.3 Zone 2 compliant - Internal and External
- Easy connection to your existing ventilation system
- Minimal setup
- High capacity refrigeration system for temperature recovery
- Requirements for airflow dependant on items stored
- Base contains legs and castors
- BioLine ventilated air distribution system
Dedicated ExGuard extraction controller and software inhibiting access to the storage chamber unless safe extraction of cabinet interior has occurred. Timing of steps in the access procedure can be adjusted to suit your specifications. Separate controller managing normal operation as with the BioPlus range.
Material: Stainless steel (Interior); White lacquered steel or stainless steel (Exterior)
Ambient range: Solid door 10/43 °C; Glass door 10/38 °C
Certifications: ATEX marking, ll 3G Ex ec nC ic IIB T1 Gc, DTI 22ATEX0249X.