Soxhlet Extraction Apparatus
Catalog # WHEA416005
Supplier: DWK Life Sciences
Specifications
- Capacity:250 ml
- Description:Soxhlet extraction apparatus with standard taper ground joints
- Pk:1
Specifications
About this item
Soxhlet extraction apparatus with standard taper ground joints is used for continuous extraction of analytes from a solid into an organic solvent.
- Manufactured from low extractable borosilicate glass
As the flask containing the solvent is heated, vapors rise in the larger outside tube, enter the water-cooled condenser and liquate and when the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask.
Certifications: Conforms to USP Type I and ASTM E 438 Type I, Class A requirements.